Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165894.1 | NP_001159366.1 | 7 - 8 | 1009 | 68 | |
NM_009652.3 | NP_033782.1 | 7 - 8 | 1009 | 68 | ||
GenBank mRNA | AK040758.1 | - | 7 - 8 | 815 | 68 | |
AK154936.1 | - | 7 - 8 | 790 | 68 | ||
BC066018.1 | - | 7 - 8 | 1009 | 5716837 | 68 | |
X65687.1 | - | 7 - 8 | 922 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |