Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291131.1 | NP_001278060.1 | 5 - 6 | 400 | 93 | |
NM_009714.3 | NP_033844.1 | 5 - 6 | 510 | 93 | ||
GenBank mRNA | AK132959.1 | - | 5 - 6 | 401 | 93 | |
BC014779.1 | - | 5 - 6 | 416 | 93 | ||
BC022106.1 | - | 5 - 6 | 472 | 5102469 | 93 | |
CB948939.1 | - | 5 - 6 | 513 | 93 | ||
U08372.1 | - | 4 - 5 | 560 | 93 | ||
U09362.1 | - | 5 - 6 | 461 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |