Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859647 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859646 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859648 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859650 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1973006 |
BC003722.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1973006 |
BQ031093.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1973006 |
NM_001171739.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1973006 |
NM_009736.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2043317 |
BC003722.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2043317 |
BC069918.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2043317 |
BC093509.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2043317 |
BQ031093.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2043317 |
AK013611.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
36287 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579798 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
109522 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
461724 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326000 |
AK144612.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326000 |
AK162291.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326000 |
BC027231.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
326000 |
NM_145972.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
43393 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
102849 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933899 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
173648 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
131011 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P119339 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116501 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2043317 |
BC003722.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2043317 |
BC069918.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2043317 |
BC093509.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2043317 |
BQ031093.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2043317 |
AK013611.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1973006 |
BC003722.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1973006 |
BQ031093.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1973006 |
NM_001171739.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1973006 |
NM_009736.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P224505 |
BC003722.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P224505 |
BC069918.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P224505 |
BC093509.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P224505 |
AK009149.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P224505 |
AK154469.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859633 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859625 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859647 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859628 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859632 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859630 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859637 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859648 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859623 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859620 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859646 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859645 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859641 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859634 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859635 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22859643 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00855889 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116501 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P119339 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P224505 |
BC003722.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P224505 |
BC069918.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P224505 |
BC093509.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P224505 |
AK009149.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P224505 |
AK154469.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104498 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116501 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P119339 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03964282 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03964266 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03964242 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03964264 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03964272 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859647 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859650 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859646 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22859648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859552 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859546 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859588 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859560 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859544 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859542 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859551 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22859572 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859628 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03964275 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859645 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859632 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03226053 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859641 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03964273 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859646 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859643 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859609 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859617 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03234052 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859630 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03418603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859601 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859599 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859598 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859637 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859619 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03652083 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859649 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859627 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859638 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03922958 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03286378 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859603 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22859614 |
- |
- |