Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291799.1 | NP_001278728.1 | 2 - 3 | 406 | 65 | |
NM_010070.4 | NP_034200.4 | 2 - 3 | 408 | 65 | ||
GenBank mRNA | AK136382.1 | - | 2 - 3 | 409 | 65 | |
AK169871.1 | - | 2 - 3 | 404 | 65 | ||
BC013066.1 | - | 2 - 3 | 402 | 3498170 | 65 | |
CJ053718.1 | - | 2 - 3 | 409 | 65 | ||
U78818.2 | - | 2 - 3 | 373 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |