Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077189.1 | NP_001070657.1 | 4 - 5 | 703 | 112 | |
NM_010187.2 | NP_034317.1 | 4 - 5 | 703 | 112 | ||
GenBank mRNA | AK080885.1 | - | 4 - 5 | 706 | 112 | |
AK089266.1 | - | 4 - 5 | 704 | 112 | ||
AK150613.1 | - | 4 - 5 | 704 | 112 | ||
AK155329.1 | - | 4 - 5 | 695 | 112 | ||
AK170035.1 | - | 4 - 5 | 693 | 112 | ||
AK171254.1 | - | 4 - 5 | 695 | 112 | ||
AK171316.1 | - | 4 - 5 | 686 | 112 | ||
BC011305.1 | - | 4 - 5 | 666 | 112 | ||
BC019758.1 | - | 4 - 5 | 666 | 112 | ||
BC038070.1 | - | 4 - 5 | 690 | 3993915 | 112 | |
M14216.1 | - | 4 - 5 | 661 | 112 | ||
M16367.1 | - | 6 - 7 | 944 | 112 | ||
M17515.1 | - | 4 - 5 | 657 | 112 | ||
U31801.1 | - | 2 - 3 | 518 | 112 | ||
U31802.1 | - | 2 - 3 | 518 | 112 | ||
U31803.1 | - | 2 - 3 | 518 | 112 | ||
U31804.1 | - | 2 - 3 | 518 | 112 | ||
U51629.1 | - | 4 - 5 | 611 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |