Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010801.3 | NP_034931.2 | 2 - 3 | 315 | 70 | |
GenBank mRNA | AF009515.1 | - | 2 - 3 | 256 | 70 | |
AF100171.1 | - | 2 - 3 | 256 | 70 | ||
AK007292.1 | - | 2 - 3 | 314 | 70 | ||
AK133448.1 | - | 2 - 3 | 316 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |