Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209748 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209745 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209749 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209747 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
531779 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
937854 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
807744 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
599302 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729037 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
127492 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
686232 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
281686 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
154736 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
782154 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
872933 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
873049 |
AK030886.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
873049 |
BC064108.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402422 |
AK007173.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402422 |
BC087878.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402422 |
NM_183103.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
206449 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
294156 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
727221 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97863 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
122379 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94927 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166047 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P143126 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209720 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209728 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209724 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209714 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209740 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209755 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209730 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209744 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209721 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209743 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209723 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209746 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209761 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209742 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209718 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209741 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209750 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209753 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209715 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209726 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209748 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209757 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209763 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209751 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209716 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209759 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209739 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22209760 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P143126 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166047 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00663494 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
AK133448.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
BC027526.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
NM_001039543.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
NM_010801.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P205106 |
AF009515.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119522 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166047 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00663494 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P143126 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03182942 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02637741 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02637732 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03182938 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P02637742 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209747 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209749 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209745 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P22209748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209716 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209695 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209702 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209776 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209700 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22209740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03083954 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209755 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02728233 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02637742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209753 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209721 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209724 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02708942 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209718 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209761 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03190280 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02637743 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03182944 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209772 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209731 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209774 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209778 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209744 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209712 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02776340 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209768 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209769 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209694 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209757 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209759 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209776 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209735 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02436321 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02575725 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209710 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02637732 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209762 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209758 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209695 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209702 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03127055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02637740 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209723 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03083953 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209742 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209741 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209716 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209771 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209748 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02998188 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209706 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209728 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209777 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209700 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209773 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209691 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209711 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209736 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209737 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209760 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03182939 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02436322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209766 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209692 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209697 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22209708 |
- |
- |