Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010929.2 | NP_035059.2 | 26 - 27 | 4885 | 73 | |
GenBank mRNA | AK143484.1 | - | 9 - 10 | 2050 | 73 | |
BC138044.1 | - | 26 - 27 | 4843 | 8861063 | 73 | |
BC145114.1 | - | 27 - 28 | 4973 | 73 | ||
M80456.1 | - | 26 - 27 | 4850 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |