Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011051.3 | NP_035181.1 | 1 - 2 | 220 | 68 | |
GenBank mRNA | AK008610.1 | - | 1 - 2 | 144 | 68 | |
AK028223.1 | - | 1 - 2 | 175 | 68 | ||
AK078044.1 | - | 1 - 2 | 148 | 68 | ||
AK147979.1 | - | 1 - 2 | 178 | 68 | ||
AK150335.1 | - | 1 - 2 | 138 | 68 | ||
BC040079.1 | - | 1 - 2 | 171 | 3156800 | 68 | |
BU604816.1 | - | 1 - 2 | 223 | 68 | ||
U49112.1 | - | 1 - 2 | 157 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |