Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291434.1 | NP_001278363.1 | 6 - 7 | 945 | 80 | |
NM_011102.4 | NP_035232.1 | 6 - 7 | 945 | 80 | ||
GenBank mRNA | AK144419.1 | - | 6 - 7 | 947 | 80 | |
BC111807.1 | - | 6 - 7 | 832 | 40039740 | 80 | |
L28035.1 | - | 6 - 7 | 1023 | 80 | ||
X67129.1 | - | 6 - 7 | 754 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |