Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011345.2 | NP_035475.1 | 9 - 10 | 1608 | 71 | |
GenBank mRNA | AK153623.1 | - | 9 - 10 | 1627 | 71 | |
BC119083.1 | - | 9 - 10 | 1559 | 8733832 | 71 | |
BC120795.1 | - | 9 - 10 | 1559 | 40129718 | 71 | |
M87862.1 | - | 9 - 10 | 1608 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |