Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272024.1 | NP_001258953.1 | 8 - 9 | 947 | 66 | |
NM_011351.2 | NP_035481.1 | 5 - 6 | 515 | 66 | ||
NR_073549.1 | 6 - 7 | 840 | 66 | |||
NR_073550.1 | 6 - 7 | 854 | 66 | |||
NR_073551.1 | 7 - 8 | 903 | 66 | |||
GenBank mRNA | AB013729.1 | - | 5 - 6 | 480 | 66 | |
AF363972.1 | - | 5 - 6 | 548 | 66 | ||
AF363973.1 | - | 5 - 6 | 548 | 66 | ||
AK031131.1 | - | 6 - 7 | 752 | 66 | ||
AK165085.1 | - | 6 - 7 | 767 | 66 | ||
BC051979.1 | - | 7 - 8 | 809 | 66 | ||
BC059007.1 | - | 8 - 9 | 862 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |