Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011491.3 | NP_035621.1 | 2 - 3 | 499 | 60 | |
GenBank mRNA | AF031035.1 | - | 2 - 3 | 499 | 60 | |
AF056244.1 | - | 2 - 3 | 346 | 60 | ||
AK002527.1 | - | 2 - 3 | 307 | 60 | ||
AK010318.1 | - | 2 - 3 | 314 | 60 | ||
BC012206.1 | - | 2 - 3 | 331 | 3665803 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |