Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 4 - 5 | 256 | 74 | |
GenBank mRNA | AF263345.1 | - | 4 - 5 | 263 | 74 | |
AK088364.1 | - | 4 - 5 | 278 | 74 | ||
AK133419.1 | - | 4 - 5 | 257 | 74 | ||
BC003477.1 | - | 4 - 5 | 246 | 74 | ||
BC008102.1 | - | 4 - 5 | 266 | 3966319 | 74 | |
D42068.1 | - | 4 - 5 | 248 | 74 | ||
U19520.1 | - | 4 - 5 | 270 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |