Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011503.4 | NP_035633.1 | 9 - 10 | 808 | 62 | |
GenBank mRNA | AF263345.1 | - | 9 - 10 | 815 | 62 | |
AK088364.1 | - | 9 - 10 | 830 | 62 | ||
AK133419.1 | - | 9 - 10 | 809 | 62 | ||
BC003477.1 | - | 9 - 10 | 798 | 62 | ||
BC008102.1 | - | 9 - 10 | 818 | 3966319 | 62 | |
D42068.1 | - | 9 - 10 | 800 | 62 | ||
U19520.1 | - | 9 - 10 | 822 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |