Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011530.3 | NP_035660.3 | 3 - 4 | 762 | 115 | |
GenBank mRNA | AK020022.1 | - | 3 - 4 | 675 | 115 | |
AK079667.1 | - | 3 - 4 | 676 | 115 | ||
AK145234.1 | - | 3 - 4 | 722 | 115 | ||
AK151376.1 | - | 3 - 4 | 672 | 115 | ||
AK151483.1 | - | 3 - 4 | 672 | 115 | ||
AK151657.1 | - | 3 - 4 | 695 | 115 | ||
AK153287.1 | - | 3 - 4 | 672 | 115 | ||
AK159200.1 | - | 3 - 4 | 728 | 115 | ||
AK161091.1 | - | 3 - 4 | 116 | |||
BC005578.1 | - | 3 - 4 | 662 | 3968225 | 115 | |
BC051257.1 | - | 3 - 4 | 675 | 6530488 | 115 | |
BC057668.1 | - | 3 - 4 | 1049 | 116 | ||
CT010387.1 | - | 2 - 3 | 609 | 115 | ||
KJ650229.1 | - | 2 - 3 | 612 | 115 | ||
M90459.1 | - | 2 - 3 | 609 | 115 | ||
U60087.1 | - | 2 - 3 | 612 | 115 | ||
U60088.1 | - | 2 - 3 | 612 | 115 | ||
U60089.1 | - | 2 - 3 | 612 | 115 | ||
U60090.1 | - | 2 - 3 | 612 | 115 | ||
U60091.1 | - | 2 - 3 | 612 | 115 | ||
U60092.1 | - | 2 - 3 | 609 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Mouse ABC Transporters 1700 Inventoried Mouse_ABC_Transporters Probe spans exons |