Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011577.1 | NP_035707.1 | 1 - 2 | 1220 | 99 | |
GenBank mRNA | AJ009862.1 | - | 1 - 2 | 1220 | 99 | |
AK144163.1 | - | 1 - 2 | 1222 | 99 | ||
BC013738.1 | - | 1 - 2 | 667 | 3586216 | 99 | |
M13177.1 | - | 1 - 2 | 705 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Mouse Chemokine Panel Mouse Tumor Metastasis Panel Mouse Immune Response Inventoried Mouse_Immune_Response Probe spans exons |