Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164078.1 | NP_001157550.1 | 10 - 11 | 1142 | 101 | |
NM_011585.4 | NP_035715.1 | 11 - 12 | 1169 | 101 | ||
GenBank mRNA | AK028340.1 | - | 9 - 10 | 809 | 101 | |
AK088486.1 | - | 11 - 12 | 1105 | 101 | ||
AK144222.1 | - | 11 - 12 | 1122 | 101 | ||
AK190403.1 | - | 2 - 3 | 227 | 101 | ||
BC027826.1 | - | 9 - 10 | 3106 | 101 | ||
BC046812.1 | - | 10 - 11 | 1025 | 5702183 | 101 | |
U00689.1 | - | 11 - 12 | 1110 | 101 | ||
U55862.1 | - | 11 - 12 | 977 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |