Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011596.5 | NP_035726.2 | 1309 | 64 | ||
GenBank mRNA | AB022323.1 | - | 1275 | 64 | ||
AF218252.1 | - | 1294 | 64 | |||
AK155055.1 | - | 1286 | 64 | |||
AK170122.1 | - | 1563 | 64 | |||
AK182287.1 | - | 1 - 2 | 77 | 64 | ||
AK195507.1 | - | 1 - 2 | 77 | 64 | ||
BC013056.1 | - | 1 - 2 | 41 | 64 | ||
BC018369.1 | - | 1262 | 64 | |||
BC108991.1 | - | 1206 | 40047832 | 64 | ||
BC108992.1 | - | 1206 | 40047833 | 64 | ||
BC112905.1 | - | 1269 | 3670722 | 64 | ||
M31226.1 | - | 1205 | 64 | |||
X55184.1 | - | 1206 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |