Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011631.1 | NP_035761.1 | 2 - 3 | 239 | 72 | |
GenBank mRNA | AK146885.1 | - | 2 - 3 | 305 | 72 | |
AK150811.1 | - | 2 - 3 | 259 | 72 | ||
AK151413.1 | - | 2 - 3 | 256 | 72 | ||
AK153550.1 | - | 2 - 3 | 259 | 72 | ||
AK159974.1 | - | 2 - 3 | 264 | 72 | ||
AK160093.1 | - | 2 - 3 | 259 | 72 | ||
AK165975.1 | - | 2 - 3 | 259 | 72 | ||
AK168952.1 | - | 2 - 3 | 259 | 72 | ||
AK169090.1 | - | 2 - 3 | 259 | 72 | ||
BC010445.1 | - | 2 - 3 | 171 | 3587178 | 72 | |
BC011439.1 | - | 2 - 3 | 245 | 3707817 | 72 | |
J03297.1 | - | 2 - 3 | 239 | 72 | ||
M16370.1 | - | 2 - 3 | 197 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |