Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290364.1 | NP_001277293.1 | 3 - 4 | 556 | 71 | |
NM_012019.3 | NP_036149.1 | 3 - 4 | 568 | 71 | ||
GenBank mRNA | AF100927.1 | - | 3 - 4 | 394 | 71 | |
AK133764.1 | - | 3 - 4 | 409 | 71 | ||
BC003292.1 | - | 3 - 4 | 431 | 3500037 | 71 | |
BC024882.1 | - | 3 - 4 | 1052 | 71 | ||
DQ016499.1 | - | 3 - 4 | 431 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |