Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253737.1 | NP_001240666.1 | 9 - 10 | 718 | 70 | |
NM_001253739.1 | NP_001240668.1 | 9 - 10 | 729 | 70 | ||
NM_001253743.1 | NP_001240672.1 | 10 - 11 | 778 | 70 | ||
NM_001253745.1 | NP_001240674.1 | 8 - 9 | 655 | 70 | ||
NM_001253746.1 | NP_001240675.1 | 8 - 9 | 655 | 70 | ||
NM_001253747.1 | NP_001240676.1 | 9 - 10 | 739 | 70 | ||
NM_013517.4 | NP_038545.1 | 10 - 11 | 859 | 70 | ||
GenBank mRNA | AK156654.1 | - | 10 - 11 | 862 | 70 | |
AK172591.1 | - | 10 - 11 | 862 | 70 | ||
AY069980.1 | - | 8 - 9 | 621 | 70 | ||
AY069981.1 | - | 8 - 9 | 621 | 70 | ||
BC119164.1 | - | 9 - 10 | 726 | 8733913 | 70 | |
BC119190.1 | - | 9 - 10 | 726 | 8733939 | 70 | |
M34163.1 | - | 10 - 11 | 861 | 70 | ||
M99371.1 | - | 10 - 11 | 851 | 70 | ||
S67072.1 | - | 9 - 10 | 710 | 70 | ||
S67073.1 | - | 8 - 9 | 590 | 70 | ||
S67074.1 | - | 9 - 10 | 629 | 70 | ||
X64223.1 | - | 9 - 10 | 718 | 70 | ||
X64224.1 | - | 9 - 10 | 729 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |