Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013622.3 | NP_038650.3 | 1 - 2 | 325 | 59 | |
GenBank mRNA | AK043873.1 | - | 1 - 2 | 326 | 59 | |
BC137958.1 | - | 1 - 2 | 252 | 8860977 | 59 | |
BC137969.1 | - | 1 - 2 | 252 | 8860988 | 59 | |
L06322.1 | - | 1 - 2 | 282 | 59 | ||
L07271.1 | - | 1 - 2 | 252 | 59 | ||
L11064.1 | - | 1 - 2 | 235 | 59 | ||
S65335.1 | - | 1 - 2 | 251 | 59 | ||
S66181.1 | - | 1 - 2 | 203 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |