Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013820.3 | NP_038848.1 | 4 - 5 | 969 | 61 | |
GenBank mRNA | AJ238540.1 | - | 4 - 5 | 912 | 61 | |
AK145936.1 | - | 4 - 5 | 950 | 61 | ||
BC054472.1 | - | 4 - 5 | 888 | 6465367 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |