Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013820.3 | NP_038848.1 | 16 - 17 | 2852 | 91 | |
GenBank mRNA | AJ238540.1 | - | 16 - 17 | 2795 | 91 | |
AK145936.1 | - | 16 - 17 | 2833 | 91 | ||
BC054472.1 | - | 16 - 17 | 2771 | 6465367 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |