Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017399.4 | NP_059095.1 | 1 - 2 | 112 | 72 | |
GenBank mRNA | AK131868.1 | - | 1 - 2 | 122 | 72 | |
BC009812.1 | - | 1 - 2 | 112 | 4159971 | 72 | |
Y14660.1 | - | 1 - 2 | 112 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |