Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017478.3 | NP_059506.1 | 1036 | 80 | ||
GenBank mRNA | AF205065.1 | - | 939 | 80 | ||
AJ251067.1 | - | 1008 | 80 | |||
AK079599.1 | - | 12 - 13 | 1061 | 80 | ||
AK088492.1 | - | 1020 | 80 | |||
AK147943.1 | - | 983 | 80 | |||
AK163668.1 | - | 11 - 12 | 1007 | 80 | ||
BC049178.1 | - | 1002 | 6309945 | 80 | ||
BC145898.1 | - | 979 | 40131201 | 80 | ||
BC145900.1 | - | 979 | 40131203 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |