Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019727.2 | NP_062701.2 | 13 - 14 | 1562 | 80 | |
GenBank mRNA | AB019214.1 | - | 13 - 14 | 1502 | 80 | |
AF154120.1 | - | 13 - 14 | 1566 | 80 | ||
AK076198.1 | - | 5 - 6 | 546 | 80 | ||
AK154079.1 | - | 13 - 14 | 1475 | 80 | ||
AK155746.1 | - | 13 - 14 | 1450 | 80 | ||
AK161241.1 | - | 13 - 14 | 1543 | 80 | ||
AK167177.1 | - | 13 - 14 | 1517 | 80 | ||
AK167993.1 | - | 13 - 14 | 1457 | 80 | ||
AK168742.1 | - | 13 - 14 | 1555 | 80 | ||
BC066044.1 | - | 13 - 14 | 1519 | 6807363 | 80 | |
BC066189.1 | - | 13 - 14 | 1453 | 30471747 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |