Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbke | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019777.3 | NP_062751.2 | 3 - 4 | 433 | 107 | |
GenBank mRNA | AB016589.1 | - | 1 - 2 | 121 | 107 | |
AK088580.1 | - | 3 - 4 | 435 | 107 | ||
AK149911.1 | - | 3 - 4 | 422 | 107 | ||
AK153046.1 | - | 3 - 4 | 434 | 107 | ||
BC037446.1 | - | 2 - 3 | 177 | 3980145 | 107 | |
CT010206.1 | - | 1 - 2 | 87 | 107 | ||
CT010313.1 | - | 1 - 2 | 87 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |