Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020332.4 | NP_065065.3 | 10 - 11 | 1592 | 69 | |
GenBank mRNA | AF001533.2 | - | 8 - 9 | 938 | 69 | |
AF274752.1 | - | 10 - 11 | 1295 | 69 | ||
AK083135.1 | - | 10 - 11 | 1593 | 69 | ||
AK086014.1 | - | 3 - 4 | 463 | 69 | ||
AK148070.1 | - | 10 - 11 | 1365 | 69 | ||
BC054379.1 | - | 10 - 11 | 1499 | 6390764 | 69 | |
DQ832285.1 | - | 10 - 11 | 1592 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |