Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950974 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950973 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950975 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
448816 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332574 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355207 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
708166 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551308 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
300890 |
AF097439.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
300890 |
AK132005.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
300890 |
BC027529.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
300890 |
NM_009749.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284626 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
231431 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
75892 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P145053 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P160467 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951023 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950982 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951004 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950974 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950996 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951036 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950976 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950994 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951000 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951037 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950991 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950998 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950987 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951027 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951029 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951024 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951026 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950993 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950979 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951034 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950986 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951031 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950997 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25951001 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950985 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25950989 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P160467 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06548191 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P145053 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC057328.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC083336.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
U89427.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
AK140753.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252410 |
BC009170.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P137568 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P106161 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P145053 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P160467 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06814315 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950975 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950973 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P25950974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950976 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950997 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950998 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951023 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950985 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951025 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950996 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950993 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951093 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950970 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950991 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950989 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951072 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25950972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951019 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25951004 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951010 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951008 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950972 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951037 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951031 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950987 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951007 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951000 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951019 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951028 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950985 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950983 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951038 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06832008 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07157579 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951034 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07157586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06814315 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950997 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950991 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06720644 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06770451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950993 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951024 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951015 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950986 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950979 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950996 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950998 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950989 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951021 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950974 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951023 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951016 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950976 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951004 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06766936 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25950980 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25951017 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07157589 |
- |
- |