Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200021170.mm.1 |
NM_023565.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021170.mm.1 |
NM_023565.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021170.mm.1 |
TC0200002577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021171.mm.1 |
NM_023565.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021171.mm.1 |
NM_023565.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021171.mm.1 |
TC0200002577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021168.mm.1 |
NM_023565.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021168.mm.1 |
NM_023565.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021168.mm.1 |
TC0200002577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021166.mm.1 |
NM_023565.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021166.mm.1 |
NM_023565.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021166.mm.1 |
TC0200002577.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200021169.mm.1 |
NM_023565.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021169.mm.1 |
NM_023565.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200021169.mm.1 |
TC0200002577.mm.1 |
MTA TC |