Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031197.2 | NP_112474.2 | 8 - 9 | 1179 | 84 | |
GenBank mRNA | AK005068.1 | - | 8 - 9 | 1180 | 84 | |
AK149532.1 | - | 1 - 2 | 292 | 84 | ||
AK165430.1 | - | 8 - 9 | 1114 | 84 | ||
BC034675.1 | - | 8 - 9 | 1098 | 4236331 | 84 | |
X15684.1 | - | 8 - 9 | 1128 | 84 | ||
X16986.1 | - | 8 - 9 | 1118 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |