Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173372.1 | NP_001166843.1 | 7 - 8 | 317 | 73 | |
NM_033648.1 | NP_387468.1 | 7 - 8 | 320 | 73 | ||
GenBank mRNA | AK018728.1 | - | 7 - 8 | 317 | 73 | |
AU041511.1 | - | 2 - 3 | 73 | |||
BC086918.1 | - | 8 - 9 | 581 | 6762406 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |