Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293313.1 | NP_001280242.1 | 8 - 9 | 999 | 68 | |
NM_138314.4 | NP_612187.2 | 8 - 9 | 1002 | 68 | ||
NM_178071.6 | NP_835172.1 | 8 - 9 | 999 | 68 | ||
GenBank mRNA | AF202048.1 | - | 7 - 8 | 903 | 68 | |
AK044962.1 | - | 8 - 9 | 973 | 68 | ||
AK085222.1 | - | 8 - 9 | 947 | 68 | ||
AK133221.1 | - | 1470 | 68 | |||
AK144915.1 | - | 8 - 9 | 1003 | 68 | ||
AK164652.1 | - | 8 - 9 | 947 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons |