Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293650.1 | NP_001280579.1 | 3 - 4 | 355 | 60 | |
NM_001293651.1 | NP_001280580.1 | 3 - 4 | 359 | 60 | ||
NM_008925.2 | NP_032951.1 | 3 - 4 | 355 | 60 | ||
GenBank mRNA | AK137661.1 | - | 3 - 4 | 322 | 60 | |
AK153931.1 | - | 3 - 4 | 330 | 60 | ||
AK159705.1 | - | 3 - 4 | 326 | 60 | ||
BC009816.1 | - | 3 - 4 | 288 | 4008182 | 60 | |
BY295975.1 | - | 3 - 4 | 356 | 60 | ||
CD807078.1 | - | 3 - 4 | 324 | 60 | ||
U92794.1 | - | 3 - 4 | 308 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |