Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293650.1 | NP_001280579.1 | 6 - 7 | 627 | 83 | |
NM_001293651.1 | NP_001280580.1 | 6 - 7 | 631 | 83 | ||
NM_008925.2 | NP_032951.1 | 6 - 7 | 627 | 83 | ||
GenBank mRNA | AK046762.1 | - | 1 - 2 | 1088 | 83 | |
AK137661.1 | - | 6 - 7 | 594 | 83 | ||
AK153931.1 | - | 6 - 7 | 602 | 83 | ||
AK159705.1 | - | 6 - 7 | 598 | 83 | ||
BC009816.1 | - | 6 - 7 | 560 | 4008182 | 83 | |
CD807078.1 | - | 6 - 7 | 596 | 83 | ||
U92794.1 | - | 6 - 7 | 580 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |