Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009484.2 | NP_033510.2 | 6 - 7 | 738 | 126 | |
GenBank mRNA | AF057367.1 | - | 6 - 7 | 718 | 126 | |
AK142344.1 | - | 8 - 9 | 826 | 126 | ||
AK142512.1 | - | 6 - 7 | 654 | 126 | ||
AK155922.1 | - | 7 - 8 | 618 | 126 | ||
AK162410.1 | - | 5 - 6 | 446 | 126 | ||
AK220463.1 | - | 7 - 8 | 826 | 126 | ||
BC053061.1 | - | 6 - 7 | 721 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |