Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163159.1 | NP_001156631.1 | 4 - 5 | 620 | 83 | |
NM_001163160.1 | NP_001156632.1 | 4 - 5 | 554 | 83 | ||
NM_009981.4 | NP_034111.1 | 4 - 5 | 456 | 83 | ||
GenBank mRNA | AK044585.1 | - | 4 - 5 | 457 | 83 | |
AK076050.1 | - | 4 - 5 | 381 | 83 | ||
AK076830.1 | - | 4 - 5 | 555 | 83 | ||
BB642590.1 | - | 4 - 5 | 457 | 83 | ||
BC018313.1 | - | 3 - 4 | 339 | 4162053 | 83 | |
L28956.1 | - | 4 - 5 | 425 | 83 | ||
Z12302.1 | - | 4 - 5 | 415 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |