Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011077.2 | NP_035207.1 | 14 - 15 | 2116 | 101 | |
GenBank mRNA | AK158481.1 | - | 14 - 15 | 2143 | 101 | |
BC137925.1 | - | 14 - 15 | 1863 | 8860944 | 101 | |
BC137926.1 | - | 14 - 15 | 1863 | 8860945 | 101 | |
EF194891.1 | - | 14 - 15 | 1650 | 101 | ||
U49908.1 | - | 14 - 15 | 2108 | 101 | ||
U73910.1 | - | 14 - 15 | 2116 | 101 | ||
U75646.1 | - | 14 - 15 | 1697 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |