Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ski | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011385.2 | NP_035515.2 | 1 - 2 | 991 | 81 | |
GenBank mRNA | AF435852.1 | - | 1 - 2 | 970 | 81 | |
BC068305.1 | - | 2 - 3 | 835 | 30093626 | 81 | |
CA451127.1 | - | 1 - 2 | 315 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons |