Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfeb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161722.1 | NP_001155194.1 | 6 - 7 | 1026 | 67 | |
NM_001161723.1 | NP_001155195.1 | 6 - 7 | 1020 | 67 | ||
NM_011549.3 | NP_035679.3 | 5 - 6 | 967 | 67 | ||
GenBank mRNA | AF079095.1 | - | 5 - 6 | 948 | 67 | |
AK078755.1 | - | 6 - 7 | 958 | 67 | ||
AK146016.1 | - | 5 - 6 | 992 | 67 | ||
AK154971.1 | - | 6 - 7 | 1024 | 67 | ||
BC064789.1 | - | 5 - 6 | 967 | 30429545 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |