Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040691.1 | NP_001035781.1 | 2 - 3 | 386 | 92 | |
NM_011677.2 | NP_035807.2 | 1 - 2 | 358 | 92 | ||
GenBank mRNA | AK146178.1 | - | 2 - 3 | 386 | 92 | |
AK168709.1 | - | 1 - 2 | 308 | 92 | ||
AK168785.1 | - | 1 - 2 | 308 | 92 | ||
BC011039.1 | - | 2 - 3 | 443 | 4009947 | 92 | |
BC052853.1 | - | 1 - 2 | 302 | 30058519 | 92 | |
U55041.1 | - | 1 - 2 | 312 | 92 | ||
X99018.1 | - | 1 - 2 | 314 | 92 | ||
Y08975.1 | - | 2 - 3 | 386 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |