Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011706.2 | NP_035836.2 | 13 - 14 | 2350 | 65 | |
GenBank mRNA | AB021665.1 | - | 13 - 14 | 2432 | 65 | |
AK008689.1 | - | 11 - 12 | 1875 | 65 | ||
AK089004.1 | - | 14 - 15 | 2365 | 65 | ||
AK209480.1 | - | 1 - 2 | 92 | 65 | ||
AK212478.1 | - | 1 - 2 | 92 | 65 | ||
BC005415.1 | - | 12 - 13 | 2297 | 3487527 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |