Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011904.3 | NP_036034.1 | 3 - 4 | 485 | 71 | |
GenBank mRNA | AF073526.1 | - | 3 - 4 | 546 | 71 | |
BC132537.1 | - | 3 - 4 | 485 | 40130814 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |