Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252547.1 | NP_001239476.1 | 7 - 8 | 1523 | 62 | |
NM_013781.3 | NP_038809.1 | 8 - 9 | 2009 | 62 | ||
NR_045535.1 | 9 - 10 | 2467 | 62 | |||
GenBank mRNA | AB030442.1 | - | 7 - 8 | 1486 | 62 | |
AB043953.1 | - | 8 - 9 | 1992 | 62 | ||
AF168364.1 | - | 8 - 9 | 1950 | 62 | ||
AK042709.1 | - | 7 - 8 | 1507 | 62 | ||
AK085885.1 | - | 1507 | 62 | |||
AK155165.1 | - | 8 - 9 | 1945 | 62 | ||
AK172072.1 | - | 9 - 10 | 2388 | 62 | ||
BC113203.1 | - | 8 - 9 | 1964 | 40048064 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |