Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013791.2 | NP_038819.1 | 5 - 6 | 545 | 64 | |
GenBank mRNA | AK037967.1 | - | 5 - 6 | 546 | 64 | |
AK077541.1 | - | 5 - 6 | 541 | 64 | ||
AK136123.1 | - | 2 - 3 | 228 | 64 | ||
AK155457.1 | - | 5 - 6 | 526 | 64 | ||
AK164822.1 | - | 5 - 6 | 541 | 64 | ||
AK165239.1 | - | 5 - 6 | 545 | 64 | ||
BC013703.1 | - | 5 - 6 | 529 | 3668925 | 64 | |
U72194.1 | - | 5 - 6 | 528 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |