Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013791.2 | NP_038819.1 | 11 - 12 | 1418 | 66 | |
GenBank mRNA | AK077541.1 | - | 11 - 12 | 1413 | 66 | |
AK136123.1 | - | 8 - 9 | 1103 | 66 | ||
BC013703.1 | - | 11 - 12 | 1402 | 3668925 | 66 | |
U72194.1 | - | 11 - 12 | 1401 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |