Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100026009.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026009.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026009.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025999.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025999.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025999.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026008.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026008.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026008.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026010.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026010.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026010.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026005.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026005.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026005.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026003.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026003.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026003.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026001.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026001.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026001.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026004.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026004.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026004.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026007.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026007.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026007.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100025998.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025998.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100025998.mm.1 |
TC0100003203.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100026000.mm.1 |
NM_013835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026000.mm.1 |
NM_013835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100026000.mm.1 |
TC0100003203.mm.1 |
MTA TC |