Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159284.1 | NP_001152756.1 | 13 - 14 | 2074 | 67 | |
NM_001284427.1 | NP_001271356.1 | 12 - 13 | 2023 | 67 | ||
NM_001284428.1 | NP_001271357.1 | 13 - 14 | 2131 | 67 | ||
NM_001284429.1 | NP_001271358.1 | 13 - 14 | 2076 | 67 | ||
NM_013870.3 | NP_038898.2 | 12 - 13 | 2023 | 67 | ||
GenBank mRNA | AF064236.1 | - | 12 - 13 | 1970 | 67 | |
AJ010305.1 | - | 12 - 13 | 1970 | 67 | ||
AK009426.1 | - | 2 - 3 | 664 | 67 | ||
AK030932.1 | - | 12 - 13 | 2024 | 67 | ||
AK083947.1 | - | 2 - 3 | 277 | 67 | ||
AK085412.1 | - | 13 - 14 | 2038 | 67 | ||
AK138898.1 | - | 13 - 14 | 2089 | 67 | ||
AK160492.1 | - | 3 - 4 | 478 | 67 | ||
AK166385.1 | - | 13 - 14 | 2091 | 67 | ||
AK170516.1 | - | 12 - 13 | 1983 | 67 | ||
BC010599.1 | - | 12 - 13 | 1964 | 3709078 | 67 | |
BC066192.1 | - | 13 - 14 | 1949 | 30433616 | 67 | |
BC069836.1 | - | 13 - 14 | 1949 | 30456568 | 67 | |
BC069839.1 | - | 13 - 14 | 1949 | 30431675 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |